5-Cyano-1H-imidazole-4-carboxamide hydrate ≥97%

Supplier: Matrix Scientific

085399-1G
102844-940EA 241.54 USD
102844-940
5-Cyano-1H-imidazole-4-carboxamide hydrate ≥97%
5-Cyano-1H-imidazole-4-carboxamide hydrate

Hazard: Irritant

Formula: C₅H₄N₄O
MW: 136.11 g/mol
MDL Number: MFCD04038014
CAS Number: 5372-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR