1H-Pyrazole ≥97%

Supplier: Matrix Scientific

Synonyms: 1H-PyrazolePyrazole

085386-100G
102844-912EA 43.08 USD
102844-912
1H-Pyrazole ≥97%
1H-Pyrazole

Hazard: Irritant/Hygroscopic

Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 186…188 °C
Melting Pt: 66…70 °C
MDL Number: MFCD00005234
CAS Number: 288-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR