5-Phenyl-3-(trifluoromethyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Phenyl-3-(trifluoromethyl)pyrazole

085367-1G
102844-868EA 383.08 USD
102844-868
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole ≥95%
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₇F₃N₂
MW: 212.18 g/mol
MDL Number: MFCD00156064
CAS Number: 4027-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR