3-(2,6-Difluorophenyl)-1H-pyrazole ≥97%

Supplier: Matrix Scientific

085361-1G
102844-852EA 623.08 USD
102844-852
3-(2,6-Difluorophenyl)-1H-pyrazole ≥97%
3-(2,6-Difluorophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₆F₂N₂
MW: 180.16 g/mol
MDL Number: MFCD03094571
CAS Number: 154258-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR