1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Phenyl-3-(trifluoromethyl)-1H-pyrazol-5(4H)-one
085357-10G
085357-1G
102844-840EA
86.15
USD
102844-840
102844-842
1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one ≥97%
1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one
Hazard: Irritant
Formula:
C₁₀H₇F₃N₂O MW: 228.17 g/mol |
MDL Number:
MFCD00051655 CAS Number: 321-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...