1-(2-Hydroxyethyl)pyrrole ≥97%

Supplier: Matrix Scientific

085320-1G
102844-736EA 476.1 USD
102844-736
1-(2-Hydroxyethyl)pyrrole ≥97%
1-(2-Hydroxyethyl)pyrrole

Hazard: Irritant

Formula: C₆H₉NO
Boiling Pt: 110…113 °C (12 Torr)
MDL Number: MFCD00191451
CAS Number: 6719-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR