3,4-Dimethyl-1,5-dihydro-2H-pyrrol-2-one ≥97%

Supplier: Matrix Scientific

085314-1G
102844-724EA 806.15 USD
102844-724
3,4-Dimethyl-1,5-dihydro-2H-pyrrol-2-one ≥97%
3,4-Dimethyl-1,5-dihydro-2H-pyrrol-2-one

Hazard: Irritant

Formula: C₆H₉NO
MW: 111.14 g/mol
MDL Number: MFCD00111570
CAS Number: 4030-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR