(5-Nitro-2-furyl)methyl aminomethanimidothioatehydrobromide ≥97%

Supplier: Matrix Scientific

085311-1G
102844-718EA 410.77 USD
102844-718
(5-Nitro-2-furyl)methyl aminomethanimidothioatehydrobromide ≥97%
(5-Nitro-2-furyl)methyl aminomethanimidothioatehydrobromide

Hazard: Irritant

Formula: C₆H₈BrN₃O₃S
MDL Number: MFCD00084866
CAS Number: 82118-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR