5-(5-Nitro-2-furyl)-1,3,4-thiadiazole-2-thiol ≥97%

Supplier: Matrix Scientific

085308-1G
102844-712EA 298.46 USD
102844-712
5-(5-Nitro-2-furyl)-1,3,4-thiadiazole-2-thiol ≥97%
5-(5-Nitro-2-furyl)-1,3,4-thiadiazole-2-thiol

Hazard: Irritant

Formula: C₆H₃N₃O₃S₂
MDL Number: MFCD00829129
CAS Number: 16865-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR