2-[(2-Furylmethyl)sulfonyl]ethanethioamide ≥97%

Supplier: Matrix Scientific

085301-1G
102844-696EA 156.92 USD
102844-696
2-[(2-Furylmethyl)sulfonyl]ethanethioamide ≥97%
2-[(2-Furylmethyl)sulfonyl]ethanethioamide

Hazard: Irritant

Formula: C₇H₉NO₃S₂
MDL Number: MFCD00085090
CAS Number: 175202-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR