Ethyl-2-nitrilo-3-(2-furyl)prop-2-enoate ≥95%

Supplier: Matrix Scientific

085296-1G
102844-680EA 553.85 USD
102844-680
Ethyl-2-nitrilo-3-(2-furyl)prop-2-enoate ≥95%
Ethyl-2-nitrilo-3-(2-furyl)prop-2-enoate

Hazard: Irritant

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD00137603
CAS Number: 23973-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR