2-Chloro-N-(thien-2-ylmethyl)acetamide ≥97%

Supplier: Matrix Scientific

085265-1G
102844-606EA 524.62 USD
102844-606
2-Chloro-N-(thien-2-ylmethyl)acetamide ≥97%
2-Chloro-N-(thien-2-ylmethyl)acetamide

Hazard: Irritant

Formula: C₇H₈ClNOS
MDL Number: MFCD00112970
CAS Number: 21403-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR