2-Ethynylthiophene ≥97%

Supplier: Matrix Scientific

085259-1G
102844-594EA 693.85 USD
102844-594
2-Ethynylthiophene ≥97%
2-Ethynylthiophene

Hazard: Irritant

Formula: C₆H₄S
MW: 108.1610 g/mol
MDL Number: MFCD03425129
CAS Number: 4298-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR