6-(1H-Imidazol-1-yl)nicotinonitrile ≥97%

Supplier: Matrix Scientific

085240-1G
102844-544EA 340 USD
102844-544
6-(1H-Imidazol-1-yl)nicotinonitrile ≥97%
6-(1H-Imidazol-1-yl)nicotinonitrile

Hazard: Irritant

Formula: C₉H₆N₄
MDL Number: MFCD08691169
CAS Number: 923156-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR