(4-Pyridinylethynyl)trimethylsilane ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(Trimethylsilyl)ethynyl]pyridine

085239-1G
102844-542EA 949.23 USD
102844-542
(4-Pyridinylethynyl)trimethylsilane ≥95%
(4-Pyridinylethynyl)trimethylsilane

Hazard: Irritant

Formula: C₁₀H₁₃NSi
MW: 175.31 g/mol
MDL Number: MFCD09258874
CAS Number: 133810-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR