4-Methylnicotinamide ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Methyl-3-pyridinecarboxamide
085232-1G
102844-526EA
920
USD
102844-526
4-Methylnicotinamide ≥97%
4-Methylnicotinamide
Hazard: Irritant
Formula:
C₇H₈N₂O MW: 136.1512 g/mol |
MDL Number:
MFCD00234322 CAS Number: 7250-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...