2,6-Dichloropyridine-4-carboximidamide hydrochloride ≥97%

Supplier: Matrix Scientific

085231-1G
102844-524EA 750.77 USD
102844-524
2,6-Dichloropyridine-4-carboximidamide hydrochloride ≥97%
2,6-Dichloropyridine-4-carboximidamide hydrochloride

Hazard: Irritant

Formula: C₆H₆Cl₃N₃
MDL Number: MFCD00052831
CAS Number: 175204-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR