6-Thien-2-ylnicotinonitrile ≥97%

Supplier: Matrix Scientific

085224-1G
102844-506EA 806.15 USD
102844-506
6-Thien-2-ylnicotinonitrile ≥97%
6-Thien-2-ylnicotinonitrile

Hazard: Irritant

Formula: C₁₀H₆N₂S
MDL Number: MFCD08741436
CAS Number: 619334-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR