tert-Butyl (3-fluoropyrid-2-yl)methylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl ((3-fluoropyridin-2-yl)methyl)carbamate

085222-1G
102844-502EA 750.77 USD
102844-502
tert-Butyl (3-fluoropyrid-2-yl)methylcarbamate ≥97%
tert-Butyl (3-fluoropyrid-2-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₅FN₂O₂
MW: 226.25 g/mol
MDL Number: MFCD08690239
CAS Number: 886851-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR