N-(1-Benzylpiperidin-4-yl)-3-nitropyridin-2-amine ≥97%

Supplier: Matrix Scientific

085203-1G
102844-450EA 949.23 USD
102844-450
N-(1-Benzylpiperidin-4-yl)-3-nitropyridin-2-amine ≥97%
N-(1-Benzylpiperidin-4-yl)-3-nitropyridin-2-amine

Hazard: Irritant

Formula: C₁₇H₂₀N₄O₂
MW: 312.37 g/mol
MDL Number: MFCD01566541
CAS Number: 185058-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR