2-Phenoxynicotinamide ≥97%

Supplier: Matrix Scientific

085166-1G
102844-372EA 184.62 USD
102844-372
2-Phenoxynicotinamide ≥97%
2-Phenoxynicotinamide

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
MDL Number: MFCD00052252
CAS Number: 111950-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR