3-Amino-6-chloropyridine-2-carboxamide ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-6-chloropicolinamide

085154-1G
102844-348EA 230.77 USD
102844-348
3-Amino-6-chloropyridine-2-carboxamide ≥97%
3-Amino-6-chloropyridine-2-carboxamide

Hazard: Irritant

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD09750671
CAS Number: 175358-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR