2-Hydroxy-3-nitro-6-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Nitro-6-(trifluoromethyl)pyridin-2(1H)-one

085152-1G
102844-342EA 523.08 USD
102844-342
2-Hydroxy-3-nitro-6-(trifluoromethyl)pyridine ≥97%
2-Hydroxy-3-nitro-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃F₃N₂O₃
MW: 208.1 g/mol
MDL Number: MFCD10696218
CAS Number: 117519-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR