2-(Cyclobutylmethoxy)-5-nitropyridine ≥97%

Supplier: Matrix Scientific

085130-1G
102844-290EA 636.92 USD
102844-290
2-(Cyclobutylmethoxy)-5-nitropyridine ≥97%
2-(Cyclobutylmethoxy)-5-nitropyridine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD11036049
CAS Number: 947534-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR