4,6-Dimethyl-3-nitropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4-Dimethyl-5-nitropyridine

085126-10G 085126-1G
102844-280EA 241.54 USD
102844-280 102844-282
4,6-Dimethyl-3-nitropyridine ≥97%
4,6-Dimethyl-3-nitropyridine

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD05982051
CAS Number: 1074-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR