4-Methyl-2-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

085100-1G
102844-220EA 1430.77 USD
102844-220
4-Methyl-2-(trifluoromethyl)pyridine ≥97%
4-Methyl-2-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₇H₆F₃N
MDL Number: MFCD07774125
CAS Number: 1620-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR