2,3,5,6-Tetrachloroisonicotinonitrile ≥97%
Supplier: Matrix Scientific
085091-1G
102844-202EA
849.23
USD
102844-202
2,3,5,6-Tetrachloroisonicotinonitrile ≥97%
2,3,5,6-Tetrachloroisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Hazard: Irritant
Formula:
C₆Cl₄N₂ |
CAS Number:
16297-06-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...