1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene ≥95%

Supplier: Matrix Scientific

Synonyms: 5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene

084072-50G
102841-450EA 517.5 USD
102841-450
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene ≥95%
1,2,3,4-Tetrachloro-5,5-dimethoxycyclopentadiene

Hazard: Irritant

Formula: C₇H₆Cl₄O₂
MW: 263.93 g/mol
MDL Number: MFCD00001353
CAS Number: 2207-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR