6-Bromo-4H-1,3-benzodioxine ≥97%

Supplier: Matrix Scientific

084047-1G
102841-394EA 806.15 USD
102841-394
6-Bromo-4H-1,3-benzodioxine ≥97%
6-Bromo-4H-1,3-benzodioxine

Hazard: Irritant

Formula: C₈H₇BrO₂
MDL Number: MFCD10703544
CAS Number: 90050-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR