4-Fluoroquinoline ≥97%

Supplier: Matrix Scientific

084034-1G
102841-362EA 1118.46 USD
102841-362
4-Fluoroquinoline ≥97%
4-Fluoroquinoline

Hazard: Irritant

Formula: C₉H₆FN
MDL Number: MFCD01685510
CAS Number: 394-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR