5,6-Dichloro-2,1,3-benzothiadiazole ≥97%

Supplier: Matrix Scientific

084024-1G
102841-338EA 383.08 USD
102841-338
5,6-Dichloro-2,1,3-benzothiadiazole ≥97%
5,6-Dichloro-2,1,3-benzothiadiazole

Hazard: Irritant

Formula: C₆H₂Cl₂N₂S
MDL Number: MFCD00277454
CAS Number: 17821-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR