5-Chloro-4-nitro-2,1,3-benzothiadiazole ≥97%

Supplier: Matrix Scientific

084023-1G
102841-336EA 402.5 USD
102841-336
5-Chloro-4-nitro-2,1,3-benzothiadiazole ≥97%
5-Chloro-4-nitro-2,1,3-benzothiadiazole

Hazard: Irritant

Formula: C₆H₂ClN₃O₂S
CAS Number: 2274-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR