2-Chloro-4,5-dimethyl-1H-benzimidazole ≥95%

Supplier: Matrix Scientific

084019-1G
102841-326EA 806.15 USD
102841-326
2-Chloro-4,5-dimethyl-1H-benzimidazole ≥95%
2-Chloro-4,5-dimethyl-1H-benzimidazole

Hazard: Irritant

Formula: C₉H₉ClN₂
MDL Number: MFCD08236705
CAS Number: 1093231-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR