2,5-Dichloro-6-fluoro-1H-benzimidazole ≥95%

Supplier: Matrix Scientific

084016-1G
102841-320EA 806.15 USD
102841-320
2,5-Dichloro-6-fluoro-1H-benzimidazole ≥95%
2,5-Dichloro-6-fluoro-1H-benzimidazole

Hazard: Irritant

Formula: C₇H₃Cl₂FN₂
MDL Number: MFCD12963758
CAS Number: 142356-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR