3-Iodo-6-methoxypyridazine ≥97%

Supplier: Matrix Scientific

083993-1G
102841-256EA 666.15 USD
102841-256
3-Iodo-6-methoxypyridazine ≥97%
3-Iodo-6-methoxypyridazine

Hazard: Irritant

Formula: C₅H₅IN₂O
MDL Number: MFCD10697533
CAS Number: 17321-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR