4-Chloro-6-phenylpyrimidine ≥97%

Supplier: Matrix Scientific

083984-1G
102841-230EA 1027.69 USD
102841-230
4-Chloro-6-phenylpyrimidine ≥97%
4-Chloro-6-phenylpyrimidine

Hazard: Irritant

Formula: C₁₀H₇ClN₂
MDL Number: MFCD00234604
CAS Number: 3435-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR