2,4-Dichloro-6-ethyl-5-fluoropyrimidine ≥97%

Supplier: Matrix Scientific

083982-1G
102841-224EA 750.77 USD
102841-224
2,4-Dichloro-6-ethyl-5-fluoropyrimidine ≥97%
2,4-Dichloro-6-ethyl-5-fluoropyrimidine

Hazard: Irritant

Formula: C₆H₅Cl₂FN₂
MDL Number: MFCD08458278
CAS Number: 137234-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR