(5-Bromopyrimidin-2-yl)piperidine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-(piperidin-1-yl)pyrimidine

083968-1G
102841-194EA 241.54 USD
102841-194
(5-Bromopyrimidin-2-yl)piperidine ≥97%
(5-Bromopyrimidin-2-yl)piperidine

Hazard: Irritant

Formula: C₉H₁₂BrN₃
MW: 242.1157 g/mol
MDL Number: MFCD00483257
CAS Number: 57356-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR