5-Bromo-4,6-dimethylpyrimidine ≥97%

Supplier: Matrix Scientific

083967-1G
102841-192EA 160 USD
102841-192
5-Bromo-4,6-dimethylpyrimidine ≥97%
5-Bromo-4,6-dimethylpyrimidine

Hazard: Irritant

Formula: C₆H₇BrN₂
MDL Number: MFCD08059561
CAS Number: 157335-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR