2-Bromo-5-(1H-pyrrol-1-yl)pyrazine ≥97%

Supplier: Matrix Scientific

083948-1G
102841-148EA 806.15 USD
102841-148
2-Bromo-5-(1H-pyrrol-1-yl)pyrazine ≥97%
2-Bromo-5-(1H-pyrrol-1-yl)pyrazine

Hazard: Irritant

Formula: C₈H₆BrN₃
MDL Number: MFCD10688563
CAS Number: 1027512-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR