1-(1-Boc-piperidino)-4-bromo-1H-pyrazole ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate
, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole
083924-1G
102841-088EA
84.62
USD
102841-088
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole ≥97%
1-(1-Boc-piperidino)-4-bromo-1H-pyrazole
Hazard: Irritant
Formula:
C₁₃H₂₀BrN₃O₂ MW: 330.23 g/mol |
MDL Number:
MFCD09831980 CAS Number: 877399-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...