5-Bromo-2-methyl-4-phenyl-1,3-oxazole ≥97%

Supplier: Matrix Scientific

083917-1G
102841-068EA 863.08 USD
102841-068
5-Bromo-2-methyl-4-phenyl-1,3-oxazole ≥97%
5-Bromo-2-methyl-4-phenyl-1,3-oxazole

Hazard: Irritant

Formula: C₁₀H₈BrNO
MDL Number: MFCD09864950
CAS Number: 20662-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR