5-Bromo-2,4-dimethyl-1,3-thiazole ≥97%

Supplier: Matrix Scientific

083914-1G
102841-058EA 75.38 USD
102841-058
5-Bromo-2,4-dimethyl-1,3-thiazole ≥97%
5-Bromo-2,4-dimethyl-1,3-thiazole

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₅H₆BrNS
Boiling Pt: 114…120 °C (15 mmHg)
MDL Number: MFCD03086099
CAS Number: 28599-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR