2,5-Dibromo-3,4-dinitrothiophene ≥97%
Supplier: Matrix Scientific
083900-1G
102841-022EA
96.92
USD
102841-022
2,5-Dibromo-3,4-dinitrothiophene ≥97%
2,5-Dibromo-3,4-dinitrothiophene
Hazard: Irritant
Formula:
C₄Br₂N₂O₄S |
MDL Number:
MFCD00015537 CAS Number: 52431-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...