3-Iodo-2-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083887-1G
102840-984EA 723.08 USD
102840-984
3-Iodo-2-(trifluoromethyl)pyridine ≥97%
3-Iodo-2-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃F₃IN
MDL Number: MFCD08234938
CAS Number: 590371-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR