5-Fluoro-2-pyrrolidin-1-ylpyridine ≥97%

Supplier: Matrix Scientific

083879-1G
102840-964EA 806.15 USD
102840-964
5-Fluoro-2-pyrrolidin-1-ylpyridine ≥97%
5-Fluoro-2-pyrrolidin-1-ylpyridine

Hazard: Irritant

Formula: C₉H₁₁FN₂
MW: 166.2 g/mol
MDL Number: MFCD18089240
CAS Number: 1287217-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR