4-Bromo-2-(1H-imidazol-1-yl)pyridine ≥97%

Supplier: Matrix Scientific

083866-1G
102840-932EA 863.08 USD
102840-932
4-Bromo-2-(1H-imidazol-1-yl)pyridine ≥97%
4-Bromo-2-(1H-imidazol-1-yl)pyridine

Hazard: Irritant

Formula: C₈H₆BrN₃
MDL Number: MFCD08668595
CAS Number: 1142194-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR