4-Bromo-2-(4-morpholino)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(4-Bromopyridin-2-yl)morpholine4-(4-Bromo-2-pyridyl)morpholine

083863-1G
102840-926EA 636.92 USD
102840-926
4-Bromo-2-(4-morpholino)pyridine ≥97%
4-Bromo-2-(4-morpholino)pyridine

Hazard: Irritant

Formula: C₉H₁₁BrN₂O
MW: 243.1004 g/mol
MDL Number: MFCD13195616
CAS Number: 1040377-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR