2-Chloro-5-(dichloromethyl)pyridine ≥97%

Supplier: Matrix Scientific

083848-1G
102840-894EA 806.15 USD
102840-894
2-Chloro-5-(dichloromethyl)pyridine ≥97%
2-Chloro-5-(dichloromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄Cl₃N
MDL Number: MFCD11505040
CAS Number: 72637-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR