4-(4-Chloropyridin-2-yl)morpholine ≥97%

Supplier: Matrix Scientific

083838-1G
102840-872EA 435.38 USD
102840-872
4-(4-Chloropyridin-2-yl)morpholine ≥97%
4-(4-Chloropyridin-2-yl)morpholine

Hazard: Irritant

Formula: C₉H₁₁ClN₂O
MDL Number: MFCD10703542
CAS Number: 1086376-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR